missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LSR Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121792
This item is not returnable.
View return policy
Description
LSR Polyclonal Antibody for Western Blot, ICC/IF
Lipolysis-stimulated lipoprotein receptor (LSR), encoded by the LSR gene, is a crucial component in lipid metabolism, particularly involved in the endocytosis of triglyceride-rich lipoproteins. It is typically expressed in liver and adipose tissues, playing a pivotal role in maintaining lipid and glucose homeostasis. LSR binds to lipoproteins in the presence of free fatty acids, facilitating the uptake and clearance of lipids from the bloodstream, which is significant in preventing atherosclerosis and other lipid-related disorders. The protein functions as a multi-subunit complex, which includes two isoforms, LSRα and LSRβ, each contributing to lipoprotein metabolism. Research has highlighted its involvement in metabolic processes and its potential impacts on obesity and insulin resistance. LSR has also been studied in the context of hepatic lipid accumulation, where its downregulation is associated with increased lipid deposition in the liver, suggesting its importance in preventing non-alcoholic fatty liver disease (NAFLD). Due to its central role in lipid metabolism, LSR has emerged as a potential therapeutic target for metabolic diseases and cardiovascular conditions.
Specifications
| LSR | |
| Polyclonal | |
| Unconjugated | |
| LSR | |
| ILDR3; immunoglobulin-like domain containing receptor 3; lipolysis stimulated lipoprotein receptor; lipolysis stimulated receptor; lipolysis-stimulated lipoprotein receptor; Lipolysis-stimulated receptor; lipolysis-stimulated remnant; lipolysis-stimulated remnant receptor; LISCH; LISCH protein; LISCH7; Liver-specific bHLH-Zip transcription factor; liver-specific bHLH-Zip transcription factor 7; liver-specific gene on mouse chromosome 7 protein; LSR | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 51599, 54135, 64355 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q86X29, Q99KG5, Q9WU74 | |
| LSR | |
| Recombinant protein Lipolysis Stimulated Lipoprotein Receptor. The antigen corresponds to amino acid range 35-205 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction