missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LSP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102815
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total LSP1.
This gene encodes an intracellular F-actin binding protein. The protein is expressed in lymphocytes, neutrophils, macrophages, and endothelium and may regulate neutrophil motility, adhesion to fibrinogen matrix proteins, and transendothelial migration. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| LSP1 | |
| Polyclonal | |
| Unconjugated | |
| LSP1 | |
| 1 BAC CH230-300K22 (Children's Hospital Oakland Research Institute) complete; 47 kDa actin binding protein; 47 kDa actin-binding protein; 52 kDa phosphoprotein; F-actin binding and cytoskeleton associated protein; leufactin (leukocyte F-actin binding protein); leukocyte specific protein 1; leukocyte-specific protein 1; lsp; LSP1; Lsp-1; lymphocyte specific 1; lymphocyte specific protein 1; lymphocyte-specific antigen WP34; Lymphocyte-specific protein 1; OTTHUMP00000014138; OTTHUMP00000069614; OTTHUMP00000069615; OTTHUMP00000069616; p50; pp52; S37; S37 protein; WP34 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16985, 361680, 4046 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P19973, P33241 | |
| LSP1 | |
| A synthesized peptide derived from human LSP1(Accession P33241), corresponding to amino acid residues W297-P339. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction