missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSP1, Mouse anti-Human, Clone: LSP1/3025, Novus Biologicals™
Mouse Monoclonal Antibody
£221.00 - £396.00
Specifications
| Antigen | LSP1 |
|---|---|
| Clone | LSP1/3025 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18282636
|
Novus Biologicals
NBP2-79725-20UG |
20 μg |
£221.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18204198
|
Novus Biologicals
NBP2-79725-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LSP1 Monoclonal specifically detects LSP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| LSP1 | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 47 kDa actin binding protein, 47 kDa actin-binding protein, 52 kDa phosphoprotein, F-actin binding and cytoskeleton associated protein, leufactin (leukocyte F-actin binding protein), leukocyte-specific protein 1, lymphocyte-specific protein 1, lymphocyte-specific protein pp52, pp52, WP34Lymphocyte-specific antigen WP34 | |
| LSP1 | |
| IgG1 κ | |
| Protein A or G purified |
| LSP1/3025 | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P33241 | |
| 4046 | |
| Recombinant fragment of human LSP1 protein (around aa174-278) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title