missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSP1 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-74048AF405
This item is not returnable.
View return policy
Description
LSP1 Polyclonal specifically detects LSP1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| LSP1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| 47 kDa actin binding protein, 47 kDa actin-binding protein, 52 kDa phosphoprotein, F-actin binding and cytoskeleton associated protein, leufactin (leukocyte F-actin binding protein), leukocyte-specific protein 1, lymphocyte-specific protein 1, lymphocyte-specific protein pp52, pp52, WP34Lymphocyte-specific antigen WP34 | |
| Rabbit | |
| 0.1 mL | |
| 4046 | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| LSP1 | |
| A synthetic peptide corresponding a C-terminal portion of the human LSP1 protein (between amino acids 250-339) [UniProt P33241]. | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction