missing translation for 'onlineSavingsMsg'
Learn More

LSM4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18374364 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374364 100 μL 100µL
18311417 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18374364 Supplier Novus Biologicals Supplier No. NBP304444100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LSM4 Polyclonal specifically detects LSM4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSM4
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias Glycine-rich protein, GRP, LSM4 homolog, U6 small nuclear RNA associated (S. cerevisiae), U6 snRNA-associated Sm-like protein LSm4, YER112W
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-139 of human LSM4 (NP_036453.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 25804
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.