missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM3 Antibody (01), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06568B
This item is not returnable.
View return policy
Description
LSM3 Monoclonal antibody specifically detects LSM3 in Human samples. It is validated for ELISA
Specifications
| LSM3 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 01 | |
| ELISA | |
| LSM3 homolog, U6 small nuclear RNA associated (S. cerevisiae), SMX4, U6 snRNA-associated Sm-like protein LSm3, USS2, YLR438C | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LSM3 (LSM3_HUMAN; Met1-Gly102). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 27258 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction