missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM2 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB29915.100uL
This item is not returnable.
View return policy
Description
Rabbit polyclonal antibody raised against partial synthetic protein of human LSM2.
- High specificity
- High reactivity
Specifications
| LSM2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial synthetic protein of human LSM2. | |
| In 1X PBS , pH 7.4 (2% sucrose, 0.09% sodium azide). | |
| C6orf28/G7b/YBL026W/snRNP | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 57819 | |
| Store at 4°C for up to one week. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:250) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Q9Y333 | |
| LSM2 | |
| A synthetic peptide corresponding to amino acids 46-95 of human LSM2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction