missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LSM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596887
This item is not returnable.
View return policy
Description
Immunogen sequence: MLFYSFFKSL VGKDVVVELK NDLSICGTLH SVDQYLNIKL TDISVTDPEK YPHMLSVKNC FIRGSVVRYV QLPADEVDTQ LLQDAARKEA LQQKQ; Positive Samples: SW620, SKOV3; Cellular Location: Nucleus.
Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family (see SNRPD2; MIM 601061). Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.[supplied by OMIM].
Specifications
| LSM2 | |
| Polyclonal | |
| Unconjugated | |
| LSM2 | |
| C6orf28; D17H6S56E2; D17H6S56E-2; Dmapl; Dmpkap; dystrophia myotonica-protein kinase associated protein; G7B; G7b alternative form; LSM2; LSM2 homolog, U6 small nuclear RNA and mRNA degradation associated; LSM2 homolog, U6 small nuclear RNA associated; LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae); LSM2 U6 small nuclear RNA and mRNA degradation associated; protein G7b; small nuclear ribonuclear protein D homolog; small ribonuclear protein G7b; SmX5; Sm-X5; snRNP; snRNP core protein SMX5; snRNP core Sm-like protein Sm-x5; U6 snRNA-associated Sm-like protein LSm2; YBL026W | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57819 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.86 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y333 | |
| LSM2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-95 of human LSM2 (NP_0670001). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur