missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM2 Antibody (1G7), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-81280AF700
This item is not returnable.
View return policy
Description
LSM2 Monoclonal specifically detects LSM2 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| LSM2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| C6orf28chromosome 6 open reading frame 28, G7b, LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae), Protein G7b, Small nuclear ribonuclear protein D homolog, snRNP, snRNP core Sm-like protein Sm-x5, U6 snRNA-associated Sm-like protein LSm2, YBL026W | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 1G7 | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| LSM2 | |
| Recombinant human LSm2 (1-95aa) purified from E. coli | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 57819 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction