missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LSM14A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578464
This item is not returnable.
View return policy
Description
Positive Control: A549, H1299, HCT116, NT2D1 Predicted Reactivity: Pig (93%), Rhesus Monkey (97%), Bovine (90%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LSM14A, Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
Specifications
| LSM14A | |
| Polyclonal | |
| Unconjugated | |
| Lsm14a | |
| 2700023B17Rik; AA407828; AlphaSNBP; AU017544; C19orf13; FAM61A; family with sequence similarity 61, member A; hRAP55; hRAP55A; LSM14 homolog A; LSM14 homolog A (SCD6, S. cerevisiae); LSM14A; LSM14A mRNA processing body assembly factor; LSM14A, mRNA processing body assembly factor; LSM14A, SCD6 homolog A; LSM14A, SCD6 homolog A (S. cerevisiae); mRAP55A; Protein FAM61A; Protein LSM14 homolog A; protein SCD6 homolog; Putative alpha-synuclein-binding protein; RAP55; RAP55A; RGD1305695; RNA-associated protein 55; RNA-associated protein 55A; Tral | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26065, 361554, 67070 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8K2F8, Q8ND56 | |
| Lsm14a | |
| Recombinant protein encompassing a sequence within the center region of human LSM14A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction