Learn More
LSM14A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531779
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCG-7. Predicted reactivity: Mouse (97%), Rat (97%), Pig (97%), Chicken (94%), Rhesus Monkey (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LSM14A, Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.Specifications
LSM14A | |
Polyclonal | |
Unconjugated | |
Lsm14a | |
2700023B17Rik; AA407828; AlphaSNBP; AU017544; C19orf13; FAM61A; family with sequence similarity 61, member A; hRAP55; hRAP55A; LSM14 homolog A; LSM14 homolog A (SCD6, S. cerevisiae); LSM14A; LSM14A mRNA processing body assembly factor; LSM14A, mRNA processing body assembly factor; LSM14A, SCD6 homolog A; LSM14A, SCD6 homolog A (S. cerevisiae); mRAP55A; Protein FAM61A; Protein LSM14 homolog A; protein SCD6 homolog; Putative alpha-synuclein-binding protein; RAP55; RAP55A; RGD1305695; RNA-associated protein 55; RNA-associated protein 55A; Tral | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26065 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8ND56 | |
Lsm14a | |
Recombinant fragment corresponding to a region within amino acids 191 and 414 of Human LSM14A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |