missing translation for 'onlineSavingsMsg'
Learn More

LSM14A Antibody, Novus Biologicals™

Product Code. 18355459 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355459 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355459 Supplier Novus Biologicals Supplier No. H00026065B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

LSM14A Polyclonal antibody specifically detects LSM14A in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSM14A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. AAH16842.1
Gene Alias alphaSNBP, C19orf13, DKFZp434D1335, FAM61A, family with sequence similarity 61, member A, hRAP55, hRAP55A, LSM14 homolog A, LSM14A, SCD6 homolog A (S. cerevisiae), Protein FAM61A, protein LSM14 homolog A, Protein SCD6 homolog, Putative alpha-synuclein-binding protein, RAP55ALSM14 homolog A (SCD6, S. cerevisiae), RAP55chromosome 19 open reading frame 13, RNA-associated protein 55, RNA-associated protein 55A
Host Species Mouse
Immunogen LSM14A (AAH16842.1, 1 a.a. - 463 a.a.) full-length human protein. MSGGTPYIGSKISLISKAEIRYEGILYTIDTENSTVALAKVRSFGTEDRPTDRPIPPRDEVFEYIIFRGSDIKDLTVCEPPKPQCSLPQDPAIVQSSLGSSTSSFQSMGSYGPFGRMPTYSQFSPSSLVGQQFGAVGVAGSSLTSFGTETSNSGTLPQSSAVGSAFTQDTRSLKTQLSQGRSSPQLDPLRKSPTMEQAVQTASAHLPAPAAVGRRSPVSTRPLPSASQKAGENQEHRRAEVHKVSRPENEQLRNDNKRQVAPGAPSAPRRGRGGHRGGRGRFGIRRDGPMKFEKDFDFESANAQFNKEEIDREFHNKLKLKEDKLEKQEKPVNGEDKGDSGVDTQNSEGNADEEDPLGPNCYYDKTKSFFDNISCDDNRERRPTWAEERRLNAETFGIPLRPNRGRGGYRGRGGLGFRGGRGRGGGRGGTFTAPRGFRGGFRGGRGGREFADFEYRKDNKVAA
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26065
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.