missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM11 Antibody (OTI1F8), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
£195.00 - £417.00
Specifications
| Antigen | LSM11 |
|---|---|
| Clone | OTI1F8 |
| Dilution | Western Blot 1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306696
|
Novus Biologicals
NBP3-45041-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306556
|
Novus Biologicals
NBP3-45041-30ul |
30 μL |
£195.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LSM11 Monoclonal antibody specifically detects LSM11 in Human,Mouse,Rat samples. It is validated for Western BlotSpecifications
| LSM11 | |
| Western Blot 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 134353 | |
| IgG2b | |
| >95% | |
| Antibody |
| OTI1F8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| 2210404M20Rik, FLJ38273, LSM11 homolog, U7 small nuclear RNA associated, LSM11 homolog, U7 small nuclear RNA associated (S. cerevisiae), LSM11, U7 small nuclear RNA associated, U7 snRNA-associated Sm-like protein LSm11 | |
| Full length human recombinant protein of human LSM11(NP_775762) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title