missing translation for 'onlineSavingsMsg'
Learn More

LSM11 Antibody (OTI1F8) [FITC], Novus Biologicals™

Product Code. 30688019 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30688019 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688019 Supplier Novus Biologicals Supplier No. NBP345041F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LSM11 Monoclonal antibody specifically detects LSM11 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSM11
Applications Western Blot
Classification Monoclonal
Clone OTI1F8
Conjugate FITC
Formulation PBS
Gene Alias 2210404M20Rik, FLJ38273, LSM11 homolog, U7 small nuclear RNA associated, LSM11 homolog, U7 small nuclear RNA associated (S. cerevisiae), LSM11, U7 small nuclear RNA associated, U7 snRNA-associated Sm-like protein LSm11
Host Species Mouse
Immunogen Full length human recombinant protein of human LSM11(NP_775762) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134353
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.