missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM1 Rabbit anti-Human, Clone: 22, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90268-50UL
This item is not returnable.
View return policy
Description
LSM1 Monoclonal specifically detects LSM1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| LSM1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 27257 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| 22 | |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
| Cancer-associated Sm-like, CASMYJL124C, LSM1 homolog, U6 small nuclear RNA associated (S. cerevisiae), Small nuclear ribonuclear CaSm, U6 snRNA-associated Sm-like protein LSm1 | |
| Recombinant Human LSM1 Protein | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction