missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LSM1 Monoclonal Antibody (4F7)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521714
This item is not returnable.
View return policy
Description
Peptide Sequence: SIDQFANLVL HQTVERIHVG KKYGDIPRGI FVVRGENVVL LGEIDLEKES DTPLQQVSIE EILEEQRVEQ QTKLEAEKLK VQALKDRGLS IPRADTLDEY.
Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
Specifications
| LSM1 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| O15116 | |
| LSM1 | |
| LSM1 (NP_055277.1, 34 a.a. ~ 133 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 4F7 | |
| Unconjugated | |
| LSM1 | |
| 2810025O06Rik; cancer-associated Sm protein; cancer-associated Sm protein; Cancer-associated Sm-like; cancer-associated Sm-like protein; CASM; Lsm1; LSM1 homolog, mRNA degradation associated; LSM1 homolog, U6 small nuclear RNA associated; LSM1 homolog, U6 small nuclear RNA associated (S. cerevisiae); LSM1 mRNA degradation associated; LSM1, U6 small nuclear RNA associated; LSM1-like protein U6 small nuclear RNA associated; small nuclear ribonuclear CaSm; U6 snRNA-associated Sm-like protein LSm1; YJL124C | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 27257 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction