missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-00249CL1
This item is not returnable.
View return policy
Description
LSM1 Polyclonal antibody specifically detects LSM1 in Human samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| LSM1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| Cancer-associated Sm-like, CASMYJL124C, LSM1 homolog, U6 small nuclear RNA associated (S. cerevisiae), Small nuclear ribonuclear CaSm, U6 snRNA-associated Sm-like protein LSm1 | |
| Produced in rabbits immunized with purified, recombinant Human LSM1 (Accession#: O15116; Met1-Tyr133) | |
| 0.1 mL | |
| Cancer | |
| 27257 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction