missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSD1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £318.00
Specifications
| Antigen | LSD1 |
|---|---|
| Dilution | Western Blot 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18385814
|
Novus Biologicals
NBP3-18640-25UL |
25 μg |
£139.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18308391
|
Bio-Techne
NBP3-18640-100UL |
100 μg |
£318.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LSD1 Polyclonal antibody specifically detects LSD1 in Human samples. It is validated for Western BlotSpecifications
| LSD1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics | |
| Whole antiserum | |
| amine oxidase (flavin containing) domain 2, AOF2lysine-specific histone demethylase 1, BHC110FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit, Flavin-containing amine oxidase domain-containing protein 2, KIAA0601KDM1, LSD1BRAF35-HDAC complex protein BHC110, lysine (K)-specific demethylase 1, lysine (K)-specific demethylase 1A, lysine-specific histone demethylase 1A | |
| Polyclonal antibody raised in rabbit against human LSD1 (Lysine-specific demethylase 1), using the full length recombinant protein. | |
| Whole Antiserum |
| Western Blot 1:1000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| O60341 | |
| 23028 | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title