missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-536
This item is not returnable.
View return policy
Description
LSD1 Polyclonal specifically detects LSD1 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation Sequencing.
Specifications
| LSD1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| amine oxidase (flavin containing) domain 2, AOF2lysine-specific histone demethylase 1, BHC110FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit, Flavin-containing amine oxidase domain-containing protein 2, KIAA0601KDM1, LSD1BRAF35-HDAC complex protein BHC110, lysine (K)-specific demethylase 1, lysine (K)-specific demethylase 1A, lysine-specific histone demethylase 1A | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NB100-536 is specific for human BHC110-KIAA0601 protein. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP Assay | |
| 1.0 mg/mL | |
| Western Blot 1:5000-1:15000, Immunohistochemistry 1:500 - 1:2000, Immunoprecipitation 2 - 5 ug/mg lysate, Immunohistochemistry-Paraffin 1:500 - 1:2000, Chromatin Immunoprecipitation (ChIP) 10 ug, Chromatin Immunoprecipitation Sequencing | |
| O60341 | |
| KDM1A | |
| The immunogen recognized by this antibody maps to a region between residues 50 and 100 of human FAD-binding protein BRAF35-HDAC complex 110 kDa using the numbering given in TrEMBL entry O60341 (GeneID 23028). | |
| 100 μL | |
| Chromatin Modifiers, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics | |
| 23028 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction