missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58483-25ul
This item is not returnable.
View return policy
Description
LSD1 Polyclonal specifically detects LSD1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| LSD1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| amine oxidase (flavin containing) domain 2, AOF2lysine-specific histone demethylase 1, BHC110FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit, Flavin-containing amine oxidase domain-containing protein 2, KIAA0601KDM1, LSD1BRAF35-HDAC complex protein BHC110, lysine (K)-specific demethylase 1, lysine (K)-specific demethylase 1A, lysine-specific histone demethylase 1A | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
| KDM1A | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VEGAAFQSRLPHDRMTSQEAACFPDIISGPQQTQKVFLFIRNRTLQLWLDNPKIQLTFEATLQQLEAPYNSDTVLVHRVHSYLERHGLINFGIY | |
| 25 μL | |
| Chromatin Modifiers, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics | |
| 23028 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction