missing translation for 'onlineSavingsMsg'
Learn More

LSD1 Antibody, Novus Biologicals™

Product Code. 18374305 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374305 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18374305 Supplier Novus Biologicals Supplier No. NBP318684

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LSD1 Polyclonal antibody specifically detects LSD1 in Human samples. It is validated for Western Blot, ChIP assay, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ChIP assay Sequencing
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSD1
Applications ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:4000, Chromatin Immunoprecipitation 1 ug/ChIP, ELISA 1:1000-1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:200-1:500, Chromatin Immunoprecipitation Sequencing 1 ug/ChIP
Formulation PBS
Gene Accession No. O60341
Gene Alias amine oxidase (flavin containing) domain 2, AOF2lysine-specific histone demethylase 1, BHC110FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit, Flavin-containing amine oxidase domain-containing protein 2, KIAA0601KDM1, LSD1BRAF35-HDAC complex protein BHC110, lysine (K)-specific demethylase 1, lysine (K)-specific demethylase 1A, lysine-specific histone demethylase 1A
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against human LSD1 (Lysine-specific demethylase 1), using a KLH-conjugated synthetic peptide from the inner part of the protein.
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 23028
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.