missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSD1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56475AF647
This item is not returnable.
View return policy
Description
LSD1 Polyclonal specifically detects LSD1 in Human, Mouse, Rat, Canine, Primate, Zebrafish samples. It is validated for Western Blot.
Specifications
| LSD1 | |
| Polyclonal | |
| Western Blot | |
| amine oxidase (flavin containing) domain 2, AOF2lysine-specific histone demethylase 1, BHC110FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit, Flavin-containing amine oxidase domain-containing protein 2, KIAA0601KDM1, LSD1BRAF35-HDAC complex protein BHC110, lysine (K)-specific demethylase 1, lysine (K)-specific demethylase 1A, lysine-specific histone demethylase 1A | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| KDM1A | |
| A portion of amino acids 100-150 of human LSD1 was used as the immunogen. | |
| 0.1 mL | |
| DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics | |
| Human, Mouse, Rat, Canine, Primate, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction