missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRWD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570121
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 85%; Dog: 92%; Guinea Pig: 85%; Horse: 92%; Human: 100%; Mouse: 85%; Pig: 92%; Rabbit: 92%; Rat: 85%; Yeast: 100%.
Required for G1/S transition. Recruits and stabilizes the origin recognition complex (ORC) onto chromatin during G1 to establish pre-replication complex (preRC) and to heterochromatic sites in post-replicated cells. Binds a combination of DNA and histone methylation repressive marks on heterochromatin. Binds histone H3 and H4 trimethylation marks H3K9me3, H3K27me3 and H4K20me3 in a cooperative manner with DNA methylation. Required for silencing of major satellite repeats. May be important ORC2, ORC3 and ORC4 stability.
Specifications
| LRWD1 | |
| Polyclonal | |
| Unconjugated | |
| LRWD1 | |
| 1200011O22Rik; AU042569; AW548074; CENP-33; Centromere protein 33; leucine rich repeats and WD repeat domain containing 1; leucine-rich repeat and WD repeat-containing protein 1; leucine-rich repeats and WD repeat domain containing 1; LOC304396; Lrwd1; ORCA; ORC-associated protein; Origin recognition complex-associated protein; similar to hypothetical protein DKFZp434K1815; testicular tissue protein Li 4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 304396 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| LRWD1 | |
| synthetic peptide directed towards the middle region of rat LRWD1. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction