missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRWD1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-03331CL1
This item is not returnable.
View return policy
Description
LRWD1 Polyclonal antibody specifically detects LRWD1 in Human,Mouse samples. It is validated for Western Blot,Immunoprecipitation (Negative)
Specifications
| LRWD1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunoprecipitation (Negative) | |
| CoraFluor 1 | |
| DKFZp434K1815, leucine-rich repeats and WD repeat domain containing 1, ORCAleucine-rich repeat and WD repeat-containing protein 1, ORC-associated protein, origin recognition complex associated, Origin recognition complex-associated protein | |
| The immunogen recognized by this antibody maps to a region between residue 432 and 482 of human leucine-rich repeats and WD repeat domain containing 1 using the numbering given in entry NP_690852.1 (GeneID 222229). | |
| 0.1 mL | |
| Cell Biology | |
| 222229 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction