missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRSAM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554527
This item is not returnable.
View return policy
Description
Immunogen sequence: REILTELEAK SETRQENYWL IQYQRLLNQK PLSLKLQEEG MERQLVALLE ELSAEHYLPI FAHHRLSLDL LSQMSPGDLA KVGVSEAGLQ HEILR Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
LRSAM1 is a multifunctional RING finger protein that selectively regulates cell adhesion molecules, has ubiquitin ligase activity, and plays a role in receptor endocytosis and viral budding.
Specifications
| LRSAM1 | |
| Polyclonal | |
| Unconjugated | |
| Lrsam1 | |
| CMT2P; E3 ubiquitin-protein ligase LRSAM1; hTAL; leucine rich repeat and sterile alpha motif containing 1; leucine-rich repeat and sterile alpha motif-containing protein 1; LRSAM1; RGD1564403; RIFLE; RING-type E3 ubiquitin transferase LRSAM1; TAL; Tsg101-associated ligase; UNQ6496/PRO21356 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90678 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UWE0 | |
| Lrsam1 | |
| Recombinant protein corresponding to Human LRSAM1. Recombinant protein control fragment (Product #RP-93015). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction