missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRSAM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38626-20ul
This item is not returnable.
View return policy
Description
LRSAM1 Polyclonal antibody specifically detects LRSAM1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| LRSAM1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| EC 6.3.2.-, FLJ31641, hTAL, leucine rich repeat and sterile alpha motif containing 1, Leucine-rich repeat and sterile alpha motif-containing protein 1, RIFLE, TALE3 ubiquitin-protein ligase LRSAM1, Tsg101-associated ligase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human LRSAM1 (NP_612370.3).,, Sequence:, MPLFFRKRKPSEEARKRLEYQMCLAKEAGADDILDISKCELSEIPFGAFATCKVLQKKVLIVHTNHLTSLLPKSCSLLSLATIKVLDLHDNQLTALPDDLGQLTALQVLNVERNQLMQLPRSIGNLTQLQTLNVKDNKLKELPDTVGELRSLRTLNISGNEIQRLPQMLAHVRTLEMLSLDASAMVYPPREVCGAGTAAILQFLCKESGLEYYPPSQYLLPILEQDGIENSRDSPDGPTDRFSREELEWQNRFSDYEKRKEQKMLEKLEFERRLELGQREHTQLLQQSSS | |
| 20 μL | |
| Lipid and Metabolism | |
| 90678 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction