missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LRRTM3 Polyclonal Antibody

Product Code. 12839242
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12839242 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12839242 Supplier Invitrogen™ Supplier No. PA521098

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is mouse brain tissue lysate. PA5-21098 can be used with blocking peptide PEP-1212.

The Leucine-rich repeat transmembrane neuronal proteins (LRRTMs) are differentially expressed in the nervous system and were recently found to instruct presynaptic and mediate postsynaptic glutamatergic differentiation, with LRRTM1 and LRRTM2 most potent at inducing presynaptic differentiation. Recent evidence suggests that LRRTM3 promotes the processing of amyloid-precursor protein (APP) by the beta-secretase BACE, leading to the proteolytic production of the A-beta42 peptide that is the main component of amyloid plaques. Furthermore, LRRTM3 maps to a region of chromosome 10 linked to both late-onset Alzheimer's disease (AD) and elevated plasma A-beta42 levels, suggesting that LRRTM3 is a functional and positional candidate gene for AD.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRTM3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene LRRTM3
Gene Accession No. D3ZAL8, Q86VH5, Q8BZ81
Gene Alias 9630044H04Rik; leucine rich repeat transmembrane neuronal 3; leucine-rich repeat transmembrane neuronal protein 3; LRRTM3; UNQ803/PRO1693
Gene Symbols LRRTM3
Host Species Rabbit
Immunogen An 18 amino acid peptide near the carboxy terminus of human LRRTM3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216028, 294380, 347731
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.