Learn More
LRRTM2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 23094-1-AP-20UL
 View more versions of this product
Product Code. 16822195
  • £135.00 / 20µL
Description
The Leucine-rich repeat transmembrane neuronal proteins (LRRTMs) are differentially expressed in the nervous system and were recently found to instruct presynaptic and mediate postsynaptic glutamatergic differentiation, with LRRTM1 and LRRTM2 most potent at inducing presynaptic differentiation. Each LRRTM protein is a type I transmembrane containing ten extracellular leucine-rich repeats and a short intracellular tail and has a developmentally regulated pattern distinct from all others. LRRTM2 interacts with PSD-95 and regulates the surface expression of AMPA receptors. LRRTM2 also functions as a neurexin ligand, binding both Neurexin 1-alpha and -beta, suggesting that LRRTM2-Neurexin1 interaction plays a critical role in regulatory excitatory synapse development.
Specifications
Specifications
LRRTM2
Polyclonal
Unconjugated
LRRTM2
KIAA0416, LRRN2, LRRTM2
Rabbit
Antigen Affinity Chromatography
RUO
107065, 26045
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43300, Q8BGA3
LRRTM2
LRRTM2 Fusion Protein Ag19422
20 μL
Primary
Human, Mouse