missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LRRTM2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12849242
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12849242 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12849242 Supplier Invitrogen™ Supplier No. PA521097

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is SK-N-SH cell lysate. PA5-21097 can be used with blocking peptide PEP-1211.

The Leucine-rich repeat transmembrane neuronal proteins (LRRTMs) are differentially expressed in the nervous system and were recently found to instruct presynaptic and mediate postsynaptic glutamatergic differentiation, with LRRTM1 and LRRTM2 most potent at inducing presynaptic differentiation. Each LRRTM protein is a type I transmembrane containing ten extracellular leucine-rich repeats and a short intracellular tail and has a developmentally regulated pattern distinct from all others. LRRTM2 interacts with PSD-95 and regulates the surface expression of AMPA receptors. LRRTM2 also functions as a neurexin ligand, binding both Neurexin 1-alpha and -beta, suggesting that LRRTM2-Neurexin1 interaction plays a critical role in regulatory excitatory synapse development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRTM2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene LRRTM2
Gene Accession No. D4A7P2, O43300, Q8BGA3
Gene Alias AI851755; BB129880; C630011A14Rik; KIAA0416; leucine rich repeat transmembrane neuronal 2; leucine-rich repeat neuronal 2 protein; leucine-rich repeat transmembrane neuronal 2 protein; leucine-rich repeat transmembrane neuronal protein 2; LRRN2; LRRTM2; mKIAA0416
Gene Symbols LRRTM2
Host Species Rabbit
Immunogen A 17 amino acid peptide near the carboxy terminus of human LRRTM2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107065, 26045, 685472
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.