missing translation for 'onlineSavingsMsg'
Learn More

LRRTM1 Antibody (OTI5H9), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18411345 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411345 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411345 Supplier Novus Biologicals Supplier No. NBP272543AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LRRTM1 Monoclonal antibody specifically detects LRRTM1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRTM1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI5H9
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias FLJ32082, leucine rich repeat transmembrane neuronal 1, leucine-rich repeat transmembrane neuronal 1 protein, leucine-rich repeat transmembrane neuronal protein 1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 182-422 of human LRRTM1 (NP_849161) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 347730
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.