missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRN3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113430
This item is not returnable.
View return policy
Description
LRRN3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Specifications
| LRRN3 | |
| Polyclonal | |
| Unconjugated | |
| LRRN3 | |
| fibronectin type III, immunoglobulin and leucine rich repeat domains 5; FIGLER5; leucine rich repeat neuronal 3; leucine rich repeat protein 3, neuronal; leucine-rich repeat neuronal protein 3; leucine-rich repeat protein, neuronal 3; Lrrn3; Nbla10363; Neuronal leucine-rich repeat protein 3; NLRR3; NLRR-3; UNQ194/PRO220 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 54674 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.196 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9H3W5 | |
| LRRN3 | |
| Recombinant Human Leucine-rich repeat neuronal protein 3 protein (201-500AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction