missing translation for 'onlineSavingsMsg'
Learn More

LRRN3/NLRR-3 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18395945 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395945 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18395945 Supplier Novus Biologicals Supplier No. NBP299841AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LRRN3/NLRR-3 Polyclonal specifically detects LRRN3/NLRR-3 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRN3/NLRR-3
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias FIGLER5, immunoglobulin and leucine rich repeat domains 5, leucine rich repeat neuronal 3, leucine-rich repeat neuronal protein 3, leucine-rich repeat protein, neuronal 3, Neuronal leucine-rich repeat protein 3, NLRR3, NLRR-3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant LRRN3/NLRR-3 (AAH35133.1; Met1-Thr628).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54674
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.