missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRN1/NLRR-1 Antibody (4G8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00057633-M09-100ug
This item is not returnable.
View return policy
Description
LRRN1/NLRR-1 Monoclonal antibody specifically detects LRRN1/NLRR-1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| LRRN1/NLRR-1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| fibronectin type III, immunoglobulin and leucine rich repeat domains 3, FIGLER3, KIAA1497, leucine rich repeat neuronal 1, leucine-rich repeat neuronal protein 1, Neuronal leucine-rich repeat protein 1, NLRR-1 | |
| LRRN1 (CAL37408.1, 1 a.a. ∽ 716 a.a) full-length recombinant protein with GST-pstS1 tag. MARMSFVIAACQLVLGLLMTSLTESSIQNSECPQLCVCEIRPWFTPQSTYREATTVDCNDLRLTRIPSNLSSDTQVLLLQSNNIAKTVDELQQLFNLTELDFSQNNFTNIKEVGLANLTQLTTLHLEENQITEMTDYCLQDLSNLQELYINHNQISTISAHAFAGLKNLLRLHLNSNKLKVIDSRWFDSTPNLEILMIGENPVIGILDMNFKPLANLRSLVLAGMYLTDIPGNALVGLDSLESLSFYDNKLVKVPQLALQKVPSLKFLDLNKNPIHKIQEGDFKNMLRLKELGINNMGELVSVDRYALDNLPELTKLEATNNPKLSYIHRLAFRSVPALESLMLNNNALNAIYQKTVESLPNLREISIHSNPLRCDCVIHWINSNKTNIRFMEPLSMFCAMPPEYKGHQVKEVLIQDSSEQCLPMISHDSFPNRLNVDIGTTVFLDCRAMAEPEPEIYWVTPIGNKITVETLSDKYKLSSEGTLEISNIQIEDSGRYTCVAQNVQGADTRVATIKVNGTLLDGTQVLKIYVKQTESHSILVSWKVNSNVMTSNLKWSSATMKIDNPHITYTARVPVDVHEYNLTHLQPSTDYEVCLTVSNIHQQTQKSCVNVTTKNAAFAVDISDQETSTALAAVMGSMFAVISLASIAVYFAKRFKRKNYHHSLKKYMQKTSSIPLNELYPPLINLWEGDSEKDKDGSADTKPTQVDTSRSYYMW | |
| 100 μg | |
| Stem Cell Markers | |
| 57633 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 4G8 | |
| Western Blot, ELISA | |
| CAL37408.1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction