missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRN1, Mouse anti-Human, Clone: 3D11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00057633-M05.100ug
This item is not returnable.
View return policy
Description
Sequence: MARMSFVIAACQLVLGLLMTSLTESSIQNSECPQLCVCEIRPWFTPQSTYREATTVDCNDLRLTRIPSNLSSDTQVLLLQSNNIAKTVDELQQLFNLTELDFSQNNFTNIKEVGLANLTQLTTLHLEENQITEMTDYCLQDLSNLQELYINHNQISTISAHAFAGLKNLLRLHLNSNKLKVIDSRWFDSTPNLEILMIGENPVIGILDMNFKPLANLRSLVLAGMYLTDIPGNALVGLDSLESLSFYDNKLVKVPQLALQKVPSLKFLDLNKNPIHKIQEGDFKNMLRLKELGINNMGELVSVDRYALDNLPELTKLEATNNPKLSYIHRLAFRSVPALESLMLNNNALNAIYQKTVESLPNLREISIHSNPLRCDCVIHWINSNKTNIRFMEPLSMFCAMPPEYKGHQVKEVLIQDSSEQCLPMISHDSFPNRLNVDIGTTVFLDCRAMAEPEPEIYWVTPIGNKITVETLSDKYKLSSEGTLEISNIQIEDSGRYTCVAQNVQGADTRVATIKVNGTLLDGTQVLKIYVKQTESHSILVSWKVNSNVMTSNLKWSSATMKIDNPHITYTARVPVDVHEYNLTHLQPSTDYEVCLTVSNIHQQTQKSCVNVTTKNAAFAVDISDQETSTALAAVMGSMFAVISLASIAVYFAKRFKRKNYHHSLKKYMQKTSSIPLNELYPPLINLWEGDSEKDKDGSADTKPTQVDTSRSYYMWSpecifications
| LRRN1 | |
| Monoclonal | |
| Unconjugated | |
| leucine rich repeat neuronal 1 | |
| FIGLER3/KIAA1497/NLRR-1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Western Blot | |
| 3D11 | |
| In 1x PBS, pH 7.4 | |
| AM392530 | |
| LRRN1 | |
| LRRN1 (CAL37408.1, 1 a.a. ∼ 716 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 57633 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction