missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Human LRRK2, GST Tag Recombinant Protein

Product Code. 10689873
Click to view available options
Quantity:
1 mg
10 μg
Unit Size:
1mg
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10689873

Brand: Thermo Scientific™ PV4873

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots; aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Related Links: LRRK2 tools for advancing Parkinson's disease research Kinase protein portfolio LanthaScreen™ Eu Kinase Binding Assay LanthaScreen™ Activity Assay Protein Form: Recombinant, Catalytic.

LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain. Mutations in LRRK2 have been associated with Parkinson's disease. LRRK2 is a member of the leucine-rich repeat kinase family and encodes a protein with an ankryin repeat region, a leucine-rich repeat domain, a kinase domain, a DFG-like motif, a RAS domain, a GTPase domain, a MLK-like domain, and a WD40 domain. The protein is present largely in the cytoplasm but also associates with the mitochondrial outer membrane.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5S007
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 2mM DTT, 50% glycerol, 150mM NaCl, 0.02% polysorbate 20, 0.5mM EDTA and no preservative; pH 7.5
Gene ID (Entrez) 120892
Molecular Weight (g/mol) 204.9 kDa
Name Human LRRK2, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 4921513O20Rik; 9330188B09Rik; augmented in rheumatoid arthritis 17; AURA17; AW561911; BC 300-268; cI-46; D630001M17Rik; DARDARIN; DKFZp434H2111; FLJ45829; Gm927; leucine rich repeat kinase 2; leucine-rich repeat kinase 2; leucine-rich repeat serine/threonine-protein kinase 2; LRR kinase 2; LRRK 2; Lrrk2; LRRK-2; PARK8; RIPK7; ROCO2
Common Name LRRK2
Gene Symbol LRRK2
KinaseFamily LRRK Kinase Family
Protein Length 970-2527
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 970-2527
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical ADP accumulation, Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Kinase Group Ser⁄Thr Kinases: TKL Group
Shipping Condition Dry Ice
System Compatibility LanthaScreen™ Antibody, LanthaScreen™ Substrate, Adapta™
Target Entry LRRK2
Target Gene Leucine-rich repeat kinase 2
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Catalytic
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction, Neurobiology
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.