missing translation for 'onlineSavingsMsg'
Learn More

LRRK2-IN-1, MedChemExpress

Product Code. 16465176
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465176 100 mg 100mg
16454238 10 mg 10mg
16440577 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16465176 Supplier Medchem Express Supplier No. HY10875

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LRRK2-IN-1 is a potent and selective LRRK2 inhibitor with IC50 of 6 nM and 13 nM for LRRK2 (G2019S) and LRRK2 (WT), respectively.

LRRK2-IN-1 is a potent and selective LRRK2 inhibitor with IC50 of 6 nM and 13 nM for LRRK2 (G2019S) and LRRK2 (WT), respectively.

Specifications

Chemical Name or Material LRRK2-IN-1
CAS 1234480-84-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C31H38N8O3
Quantity 100 mg
Solubility Information DMSO : 30 mg/mL (52.57 mM; Need ultrasonic)
SMILES O=C(C1=CC(OC)=C(NC2=NC=C3C(N(C4=C(C(N3C)=O)C=CC=C4)C)=N2)C=C1)N5CCC(CC5)N6CCN(CC6)C
Molecular Weight (g/mol) 570.69
Formula Weight 570.69
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.