missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LRRK2 [G2019S], GST Tag Recombinant Protein
LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain.
£439.00 - £12060.00
Specifications
Accession Number | NP_940980.2, G2019S |
---|---|
Gene Alias | LRRK2 G2019S |
KinaseFamily | LRRK Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10499963
|
Invitrogen
PV4881 |
10μg |
£439.00
Each |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15468683
|
Invitrogen
PR8764C |
100μg |
£2535.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15448773
|
Invitrogen
PV4882 |
1mg |
£12060.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Mutations in the LRRK2 gene have been associated with Parkinson's disease. The G2019S mutation of LRRK2 is associated with an increase in catalytic activity.
Specifications
NP_940980.2, G2019S | |
LRRK Kinase Family | |
Recombinant | |
Baculovirus |
LRRK2 G2019S | |
Human | |
GST Tag |