missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LRRK2 [D1994A], GST Tag Recombinant Protein

Product Code. 15498803
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
This item is not returnable. View return policy

Product Code. 15498803

Brand: Invitrogen™ PV6052

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain. The protein is present largely in the cytoplasm but also associates with the mitochondrial outer membrane. Mutations in LRRK2 (R1441C Mutant) have been associated with Parkinson disease-8.
TRUSTED_SUSTAINABILITY

Specifications

Concentration See Label
For Use With (Application) Kinase Assay
Formulation proprietary buffer with no preservative
Molecular Weight (g/mol) 205 kDa
Name Human LRRK2 [D1994A], GST Tag
Quantity 1 mg
Regulatory Status RUO - research use only
Common Name LRRK2 (D1994A Mutant)
KinaseFamily LRRK Kinase Family
Protein Length 970-2527; D1994A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.