missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LRRK2 [D1994A], GST Tag Recombinant Protein
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
Description
LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain. The protein is present largely in the cytoplasm but also associates with the mitochondrial outer membrane. Mutations in LRRK2 (R1441C Mutant) have been associated with Parkinson disease-8.
Specifications
Specifications
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | proprietary buffer with no preservative |
| Molecular Weight (g/mol) | 205 kDa |
| Name | Human LRRK2 [D1994A], GST Tag |
| Quantity | 100 μg |
| Regulatory Status | RUO - research use only |
| Common Name | LRRK2 (D1994A Mutant) |
| KinaseFamily | LRRK Kinase Family |
| Protein Length | 970-2527; D1994A |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction