missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LRRK2 Antibody Packantibody specifically detects LRRK2 in Human, Mouse, Bovine, C. elegans, Insect, Plant, Human (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow (Intracellular), Gene Knock-Out
Specifications
Specifications
| Antigen | LRRK2 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Conjugate | Unconjugated |
| Gene Alias | augmented in rheumatoid arthritis 17, AURA17, Dardarin, DKFZp434H2111, EC 2.7.11.1, FLJ45829, leucine-rich repeat kinase 2, leucine-rich repeat serine/threonine-protein kinase 2, PARK8DARDARIN, Parkinson disease (autosomal dominant) 8, RIPK7, ROCO2 |
| Immunogen | See individual datasheets. |
| Quantity | 1 Pack |
| Regulatory Status | RUO |
| Research Discipline | Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase |
| Gene ID (Entrez) | 120892 |
| Target Species | Human, Mouse, Bovine, C. elegans, Insect |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?