missing translation for 'onlineSavingsMsg'
Learn More

LRRK2 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB110-58771MFV610

Product Code. 18773283

  • £486.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LRRK2 Polyclonal antibody specifically detects LRRK2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow (Cell Surface), KnockDown, KnockDown
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LRRK2
Polyclonal
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Knockout Validated, Knockdown Validated
augmented in rheumatoid arthritis 17, AURA17, Dardarin, DKFZp434H2111, EC 2.7.11.1, FLJ45829, leucine-rich repeat kinase 2, leucine-rich repeat serine/threonine-protein kinase 2, PARK8DARDARIN, Parkinson disease (autosomal dominant) 8, RIPK7, ROCO2
A synthetic peptide made to a C-terminal region, within residues 2500-2527 of the human LRRK2 protein, conjugated to KLH. [Swiss-Prot# Q5S007]
0.1 mL
Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase
120892
Store at 4C in the dark.
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
mFluor Violet 610 SE
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse, Rat, Primate
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.