missing translation for 'onlineSavingsMsg'
Learn More

LRRK1 Antibody (3G8), Novus Biologicals™

Product Code. 18429689 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429689 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429689 Supplier Novus Biologicals Supplier No. H00079705M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LRRK1 Monoclonal antibody specifically detects LRRK1 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRK1
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Sandwich ELISA
Classification Monoclonal
Clone 3G8
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH05408
Gene Alias EC 2.7.11.1, FLJ23119, KIAA1790FLJ27465, leucine-rich repeat kinase 1, leucine-rich repeat serine/threonine-protein kinase 1, RIPK6, Roco1
Host Species Mouse
Immunogen LRRK1 (AAH05408, 560 a.a. ∽ 659 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.PGAASDRSEHDLTPMDGETFSQHLQAVKILAVRDLIWVPRRGGDVIVIGLEKDSEAQRGRVIAVLKARELTPHGIMPVSSVKVCWAGWPVRDMVYMAAVM
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 79705
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.