missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRFIP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27199SS
This item is not returnable.
View return policy
Description
LRRFIP2 Polyclonal specifically detects LRRFIP2 in Human, Equine, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| LRRFIP2 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| DKFZp434H2035, FLJ20248, FLJ22683, FLJ58304, HUFI-2, leucine rich repeat (in FLII) interacting protein 2, leucine-rich repeat flightless-interacting protein 2, LRR FLII-interacting protein 2 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 9209 | |
| Human, Equine, Primate | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Immunohistochemistry-Paraffin 5ug/ml | |
| Q9Y608 | |
| LRRFIP2 | |
| Partial synthetic peptide between amino acids 150-210 of human LRRFIP2. | |
| 0.025 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (CRA_a, 721 amino acids, CRA_b, 742 amino acids), monkey and horse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction