missing translation for 'onlineSavingsMsg'
Learn More

LRRC8B Antibody, DyLight 550, Novus Biologicals™

Product Code. 18368983 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368983 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368983 Supplier Novus Biologicals Supplier No. NBP299221R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LRRC8B Polyclonal specifically detects LRRC8B in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC8B
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias KIAA0231TA-LRRPleucine-rich repeat-containing protein 8B, leucine rich repeat containing 8 family, member B, MGC42220, T cell activation leucine repeat rich protein, TALRRP, T-cell activation leucine repeat-rich protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human LRRC8B.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 23507
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.