missing translation for 'onlineSavingsMsg'
Learn More

LRRC7 Antibody - Azide and BSA Free, R&D Systems™

Product Code. 18666686 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18666686 20 μg 20µL
18643219 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18666686 Leverandør Novus Biologicals Leverandørnr. NBP30564320ul
 Restricted Item
Estimated Shipment: 03-12-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

LRRC7 Polyclonal specifically detects LRRC7 in Human/Mouse/Rat samples. It is validated for Immunocytochemistry/Immunofluorescence/Western Blot.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen LRRC7
Applications Immunocytochemistry/Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias DKFZp686I1147, leucine rich repeat containing 7, MGC144918
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 730-890 of human LRRC7 (NP_065845.1). FPQPLDSKPLLSQREAVPPGNIPQRPDRLPMSDTFTDNWTDGSHYDNTGFVAEETTAENANSNPLLSSKSRSTSSHGRRPLIRQDRIVGVPLELEQSTHRHTPETEVPPSNPWQNWTRTPSPFEDRTAFPSKLETTPTTSPLPERKEHIKESTEIPSPFSP
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57554
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.