missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRC6 Monoclonal Antibody (OTI7E9)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526778
This item is not returnable.
View return policy
Description
LRRC6 Monoclonal Antibody for Western Blot, IHC (P)
LRRC6 may be involved in spermatocytogenesis or prophase of meiosis.
Specifications
| LRRC6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86X45 | |
| LRRC6 | |
| Full length human recombinant protein of LRRC6 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7E9 | |
| Unconjugated | |
| LRRC6 | |
| CILD19; leucine rich repeat containing 6; leucine rich repeat containing 6 (testis); leucine-rich repeat-containing 6 (testis); leucine-rich repeat-containing protein 6; leucine-rich repeat-containing protein 6; protein tilB homolog; leucine-rich testis-specific protein; LRRC6; Lrtp; Mc2; Protein tilB homolog; testis-specific leucine-rich repeat protein; Tslrp; Unknown (protein for MGC:138048) | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23639 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction