Learn More
LRRC6 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26400-1-AP-20UL
This item is not returnable.
View return policy
Description
The LRRC6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LRRC6. This antibody recognizes human, mouse antigen. The LRRC6 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| LRRC6 | |
| 450 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, ELISA | |
| LRRC6, LRTP, TSLRP | |
| 20 μL | |
| 23639 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |