missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC50 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03754-100UL
This item is not returnable.
View return policy
Description
LRRC50 Polyclonal specifically detects LRRC50 in Rat samples. It is validated for Western Blot.
Specifications
| LRRC50 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 123872 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| CILD13, DKFZp434A119, FLJ25330, leucine rich repeat containing 50, ODA7leucine-rich repeat-containing protein 50, outer row dynein assembly 7 homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 516-725 of human LRRC50 (NP_848547.4). | |
| 100 μL | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction