missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC50 Antibody (OTI2C4), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72536FR
This item is not returnable.
View return policy
Description
LRRC50 Monoclonal specifically detects LRRC50 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| LRRC50 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| DNAAF1 | |
| Full length recombinant protein of human LRRC50 (NP_848547) produced in HEK293T cells. | |
| 0.1 mL | |
| 123872.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| OTI2C4 | |
| Western Blot, Flow Cytometry | |
| CILD13, DKFZp434A119, FLJ25330, leucine rich repeat containing 50, ODA7leucine-rich repeat-containing protein 50, outer row dynein assembly 7 homolog | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction