missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC4C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5111335
This item is not returnable.
View return policy
Description
Immunogen sequence: ATTTPFSYFS TVTVETMEPS QDEARTTDNN VGPTPVVDWE TTNVTTSLTP QSTRSTEKTF TIPVTDINSG IP
NGL1 is a specific binding partner for netrin G1 (NTNG1), which is a member of the netrin family of axon guidance molecules (Lin et al., 2003).
Specifications
| LRRC4C | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HCJ2 | |
| LRRC4C | |
| Recombinant Protein corresonding to Human LRRC4C | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry | |
| Unconjugated | |
| LRRC4C | |
| 6430556C10Rik; KIAA1580; Leucine-rich repeat-containing protein 4C; LRRC4C; Netrin-G1 ligand; Ngl1; NGL-1; RGD1311013; UNQ292/PRO331 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57689 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction